(+)-Quinidine, Thermo Scientific Chemicals
(+)-Quinidine, Thermo Scientific Chemicals
(+)-Quinidine, Thermo Scientific Chemicals
(+)-Quinidine, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(+)-Quinidine, Thermo Scientific Chemicals
CAS: 56-54-2 | C20H24N2O2 | 324.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFA12559.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7758-05-6
IUPAC Namepotassium iodate
Molecular FormulaIKO3
InChI KeyJLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES[K+].[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Iodometric Titration)≥97.5 to ≤102.5%
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White
Loss on Drying<1.0% (200?C, constant weight)
(+)-Quinidine is used to treat or prevent many types of irregular heartbeats (heart arrhythmias such as atrial fibrillation). It may also be used to treat malaria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in methanol, chloroform, alcohol (1 g/36ml), benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Dan M.Roden, M.D.; Raymond L.Woosley, M.D., Ph.D.; R.Kirby Primm, M.D. Incidence and clinical features of the quinidine-associated long QT syndrome: Implications for patient care. American Heart Journal. 1986, 111, (6)1088-1093
  2. Alain Borgeat, MD; Jean-Jacques Goy, MD; Raymond Maendly, MD; Urs Kaufmann, MD; MilanGrbic, MD; Ueli Sigwart, MD. Flecainide versus quinidine for conversion of atrial fibrillation to sinus rhythm. The American Journal of Cardiology. 1986, 58, (6)496-498