Phenoxyacetic acid, 98%, Thermo Scientific Chemicals
Phenoxyacetic acid, 98%, Thermo Scientific Chemicals
Phenoxyacetic acid, 98%, Thermo Scientific Chemicals
Phenoxyacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenoxyacetic acid, 98%, Thermo Scientific Chemicals
CAS: 122-59-8 | C8H8O3 | 152.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12541.22100 g
Catalog number ALFA12541.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenoxyacetic acid
CAS122-59-8
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Phenoxyacetic acid is a flavoring ingredient.�Its photodegradation using titanium dioxide as photocatalyst has been studied. Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenoxyacetic acid is a flavoring ingredient. Its photodegradation using titanium dioxide as photocatalyst has been studied. Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only