3-Acetylcoumarin, 98+%, Thermo Scientific Chemicals
3-Acetylcoumarin, 98+%, Thermo Scientific Chemicals
3-Acetylcoumarin, 98+%, Thermo Scientific Chemicals
3-Acetylcoumarin, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Acetylcoumarin, 98+%, Thermo Scientific Chemicals
CAS: 3949-36-8 | C11H8O3 | 188.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA12532.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1070-83-3
IUPAC Name3,3-dimethylbutanoic acid
Molecular FormulaC6H12O2
InChI KeyMLMQPDHYNJCQAO-UHFFFAOYSA-N
SMILESCC(C)(C)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%