Phenothiazine, 98+%, Thermo Scientific Chemicals
Phenothiazine, 98+%, Thermo Scientific Chemicals
Phenothiazine, 98+%, Thermo Scientific Chemicals
Phenothiazine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenothiazine, 98+%, Thermo Scientific Chemicals
CAS: 92-84-2 | C12H9NS | 199.271 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12517.0I5000 g
Catalog number ALFA12517.0I
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5000 g
Request bulk or custom format
Chemical Identifiers
CAS5989-54-8
IUPAC Name(4S)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Molecular FormulaC10H16
InChI KeyXMGQYMWWDOXHJM-SNVBAGLBSA-N
SMILESCC(=C)[C@H]1CCC(C)=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation(20°C, 589 nm) (neat) =< -60°
Appearance (Form)Liquid
Refractive index1.4690 to 1.4760 (20°C, 589 nm)
GC>=95 %
Appearance (Color)Clear colorless to light yellow
View more
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
Solubility
Soluble in water (0.13 mg/ml at 23°C), ethanol, acetone, and ether. Insoluble in chloroform.
Notes
Store under dry inert gas. Protect from sunlight. Ensure good ventilation.
RUO – Research Use Only
General References:
  1. A S Horn.; S H Snyder. Chlorpromazine and dopamine: conformational similarities that correlate with the antischizophrenic activity of phenothiazine drugs. PNAS. 1971, 68 (10),2325-2328.
  2. Lea Assed Bezerra Silva et. al. Antimicrobial Photodynamic Therapy for the Treatment of Teeth with Apical Periodontitis: A Histopathological Evaluation. Journal of Endodontics. 2012, 38 (3),360-366.
  3. Lithiation in THF gives orange crystals of (phenothiazin-10-yl)lithium.3THF. Carboxylation and the orientation of a second lithiation have been studied: Angew. Chem. Int. Ed., 34, 921 (1995).