Glycolic acid, 98%, Thermo Scientific Chemicals
Glycolic acid, 98%, Thermo Scientific Chemicals
Glycolic acid, 98%, Thermo Scientific Chemicals
Glycolic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycolic acid, 98%, Thermo Scientific Chemicals
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA12511.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS123-46-6
IUPAC Name[(hydrazinecarbonyl)methyl]trimethylazanium chloride
Molecular FormulaC5H14ClN3O
InChI KeyYSULOORXQBDPCU-UHFFFAOYSA-N
SMILES[Cl-].C[N+](C)(C)CC(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration Argentometric>=98.5 %
Appearance (Form)Crystalline powder
Melting point187°C to 192°C
Glycolic acid is used for acne and anti-aging treatment. It is also employed in the textile industry as a dyeing and tanning agent. It is used in emulsion polymers, solvents and additives for ink and paints. It is also useful as an intermediate for organic synthesis such as redox reactions, esterification and long chain polymerization. It acts as a monomer in the preparation of polyglycolic acid and other biocompatible copolymers. Further, it is used as a preservative and flavoring agent in organic synthesis. It is also used in adhesives and plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycolic acid is used for acne and anti-aging treatment. It is also employed in the textile industry as a dyeing and tanning agent. It is used in emulsion polymers, solvents and additives for ink and paints. It is also useful as an intermediate for organic synthesis such as redox reactions, esterification and long chain polymerization. It acts as a monomer in the preparation of polyglycolic acid and other biocompatible copolymers. Further, it is used as a preservative and flavoring agent in organic synthesis. It is also used in adhesives and plastics.
Solubility
Soluble in water, methanol, diethyl ether, acetone, acetic acid and ethyl acetate.
Notes
Hygroscopic. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Datta, D.; Kumar, S. Modeling Using Response Surface Methodology And Optimization Using Differential Evolution Of Reactive Extraction Of Glycolic Acid. Chem. Eng. Commun. 2015, 202 (1), 59-69.
  2. Steward, K. M.; Corbett, M. T.; Goodman, C. G.; Johnson, J. S. Correction to Asymmetric Synthesis of Diverse Glycolic Acid Scaffolds via Dynamic Kinetic Resolution of α-Keto Esters. J. Am. Chem. Soc. 2015, 137 (11), 3991-3991.