Piperidine, 99%, Thermo Scientific Chemicals
Piperidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Piperidine, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA12442.AE100 mL
Catalog number ALFA12442.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS3202-33-3
IUPAC Name4-phenoxypiperidine
Molecular FormulaC11H15NO
InChI KeyKBYPITRKIJKGMD-UHFFFAOYSA-N
SMILESC1CC(CCN1)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
FormPowder or crystals or crystalline powder
Appearance (Color)Brown
Assay from Supplier's CofA≥97.5% (GC)
Extensively finds applications as a base, a nucleophile and a solvent. Used for the production of dipiperidinyl dithiuram tetrasulfide, which is used as a rubber vulcanization accelerator. It is commonly used in solid phase peptide synthesis (F-MOC Chemistry) and in chemical degradation reactions (sequencing of DNA). Used in making rubber, curing epoxy resins and in fine chemicals, and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.