1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
CAS: 629-11-8 | C6H14O2 | 118.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Catalog number ALFA12439.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS90774-69-9
IUPAC Namehydrogen 4-(trifluoromethyl)aniline chloride
Molecular FormulaC7H7ClF3N
InChI KeyCZGDYZDUMJDQSD-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)White
FormPowder
1,6-Hexanediol is used in polymer synthesis such as polyester, polyurethane and nylon. It is used as an intermediate to adhesives, acrylics and dyestuffs. Further, it is employed in gasoline refining and pharmaceutical production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,6-Hexanediol is used in polymer synthesis such as polyester, polyurethane and nylon. It is used as an intermediate to adhesives, acrylics and dyestuffs. Further, it is employed in gasoline refining and pharmaceutical production.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
RUO – Research Use Only
General References:
  1. Xie, J.; Falcone, D. D.; Davis, R. J. Restructuring of supported PtSn bimetallic catalysts during aqueous phase oxidation of 1,6-hexanediol. J. Catal. 2015, 332, 38-50.
  2. Bornadel, A.; Hatti-Kaul, R.; Hollmann, F.; Kara, S. A Bi-enzymatic Convergent Cascade for ε-Caprolactone Synthesis Employing 1,6-Hexanediol as a ‘Double-Smart Cosubstrate’. ChemCatChem 2016, 7 (16), 2442-2445.