Sodium chloroacetate, 98%, Thermo Scientific Chemicals
Sodium chloroacetate, 98%, Thermo Scientific Chemicals
Sodium chloroacetate, 98%, Thermo Scientific Chemicals
Sodium chloroacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium chloroacetate, 98%, Thermo Scientific Chemicals
CAS: 3926-62-3 | C2H2ClNaO2 | 116.476 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
250 g
500 g
Catalog number ALFA12379.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium chloroacetate
CAS3926-62-3
Health Hazard 1H301-H315
Health Hazard 2GHS H Statement
H301-H315
Toxic if swallowed.
Causes skin irritation.
Health Hazard 3P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
View more
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
Solubility
Soluble in water, ether, chloroform, benzene and alcohol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. A convenient method for the conversion of arylamines to the corresponding N,N-diacetic acids is by lithiation followed by reaction with sodium chloroacetate. For discussion and limitations, see: Synthesis, 548 (1989).
  2. Ding, F.; Qian, X.; Zhang, Q.; Wu, H.; Liu, Y.; Xiao, L.; Deng, H.; Du, Y.; Shi, X. Electrochemically induced reversible formation of carboxymethyl chitin hydrogel and tunable protein release. New J. Chem. 2015, 39 (2), 1253-1259.
  3. Zhang, A.; Cheng, L.; Hong, S.; Yang, C.; Lin, Y. Preparation of anti-fouling silicone elastomers by covalent immobilization of carboxybetaine. RSC Adv. 2015, 5 (107), 88456-88463.