Urea, 98+%, Thermo Scientific Chemicals
Urea, 98+%, Thermo Scientific Chemicals
Urea, 98+%, Thermo Scientific Chemicals
Urea, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea, 98+%, Thermo Scientific Chemicals
CAS: 57-13-6 | CH4N2O | 60.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
Catalog number ALFA12360.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Thiosulphate (S2O3)=<100 ppm
Heavy metals (as Pb)=<10 ppm
Iodate (IO3)=<4 ppm
Appearance (Color)White
Titration Argentometric98.5 to 101.5 %
View more
Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.
Solubility
Soluble in water, alcohol, methanol, glycerol, benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.
Notes
Stable at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Widely used in diazotization reactions for the destruction of excess HNO2.
  2. For the one-pot conversion of carboxylic acids to nitriles, by heating with urea in the presence of 1.5-2.0 moles of sulfamic acid, see: Chimia, 25, 94 (1971).
  3. For a procedure for conversion of carboxylic acids to primary amides by heating with urea, see: Org. Synth. Coll., 3, 768 (1955); 4, 513 (1963).
  4. Reacts with alcohols on prolonged heating to give alkyl carbamates; see, e.g.: Org. Synth. Coll., 1, 140 (1941).
  5. Zhang, Y.; Han, C.; Nadagouda, M. N.; Dionysiou, D. D. The fabrication of innovative single crystal N, F-codoped titanium dioxide nanowires with enhanced photocatalytic activity for degradation of atrazine. Appl. Catal. B 2015, 168-169, 550-558.
  6. Amendola, V.; Fabbrizzi, L.; Mosca, L. Anion recognition by hydrogen bonding: urea-based receptors. Chem. Soc. Rev. 2010, 39, 3889-3915.