Sodium glycolate, 97%, Thermo Scientific Chemicals
Sodium glycolate, 97%, Thermo Scientific Chemicals
Sodium glycolate, 97%, Thermo Scientific Chemicals
Sodium glycolate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium glycolate, 97%, Thermo Scientific Chemicals
CAS: 2836-32-0 | C2H3NaO3 | 98.033 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA12341.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS87-69-4
IUPAC Name2,3-dihydroxybutanedioic acid
Molecular FormulaC4H6O6
InChI KeyFEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILESOC(C(O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to almost white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point168°C to 172°C
Loss on drying=<0.5 % (1 g, 105°C)
View more
Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Dragan, F.; Kacso, I.; Dreve, S.; Martin, F.; Borodi, G.; Bratu, I.; Earar, K. Compatibility Study of Ibuprofen with Some Excipients Employed for Solid Dosage Forms. Rev. Chim. 2015, 66 (2), 191-195.
  2. Dai, W.; Pan, X.; Chen, S.; Chen, C.; Chen, W.; Zhang, H.; Ye, Z. ZnO homojunction UV photodetector based on solution-grown Sb-doped p-type ZnO nanorods and pure n-type ZnO nanorods. RSC Adv. 2015, 5 (9), 6311-6314.