Iodine, 99+%, Thermo Scientific Chemicals
Iodine, 99+%, Thermo Scientific Chemicals
Iodine, 99+%, Thermo Scientific Chemicals
Iodine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodine, 99+%, Thermo Scientific Chemicals
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA12278.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS16423-68-0
IUPAC Namedisodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6I4Na2O5
InChI KeyIINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to brown
Infrared spectrumConforms
UV Lambda max524 to 527 nm
Appearance (Form)Powder
BSC CertificationPasses