Iodine, 99+%, Thermo Scientific Chemicals
Iodine, 99+%, Thermo Scientific Chemicals
Iodine, 99+%, Thermo Scientific Chemicals
Iodine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodine, 99+%, Thermo Scientific Chemicals
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
50 g
250 g
Catalog number ALFA12278.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS10558-25-5
IUPAC Name4-bromo-3,5-dimethyl-1,2-oxazole
Molecular FormulaC5H6BrNO
InChI KeyGYHZPSUAMYIFQD-UHFFFAOYSA-N
SMILESCC1=C(Br)C(C)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4865-1.4915 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.