Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals
Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals
Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals
Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals
CAS: 6132-04-3 | C6H9Na3O9 | 294.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
10,000 g
Catalog number ALFA12274.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS39389-20-3
IUPAC Name1,2-diethenylbenzene; 2-ethenylbenzene-1-sulfonic acid
Molecular FormulaC18H18O3S
InChI KeySIWVGXQOXWGJCI-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1C=C.OS(=O)(=O)C1=CC=CC=C1C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Exchange capacityTotal capacity >/= 4.70 eq/Kg
Residual water< / = 1.6%
Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Stable and incompatible with bases, reducing agents, oxidizing agents.
RUO – Research Use Only
Chutipongtanate, S.; Thongboonkerd, V. Establishment of a novel colorimetric assay for high-throughput analysis of calcium oxalate crystal growth modulation. Analyst 2010, 135, 1309-1314.