Benzhydrazide, 98%, Thermo Scientific Chemicals
Benzhydrazide, 98%, Thermo Scientific Chemicals
Benzhydrazide, 98%, Thermo Scientific Chemicals
Benzhydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzhydrazide, 98%, Thermo Scientific Chemicals
CAS: 613-94-5 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA12268.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS536-74-3
IUPAC Nameethynylbenzene
Molecular FormulaC8H6
InChI KeyUEXCJVNBTNXOEH-UHFFFAOYSA-N
SMILESC#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5470 to 1.5500 (20°C, 589 nm)