Pyridine-2,6-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Pyridine-2,6-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Pyridine-2,6-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Pyridine-2,6-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine-2,6-dicarboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 499-83-2 | C7H5NO4 | 167.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12263.1425 g
Catalog number ALFA12263.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7646-78-8
IUPAC Nametin(IV) chloride
Molecular FormulaCl4H4Sn
InChI KeyHTGSJGUTBKFNJC-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration Argentometric0.95 to 1.10 M
Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.
Solubility
Insoluble in water.
Notes
Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only
General References:
  1. Ge, J.; Yang, Q.; Xie, G.; Chen, S.; Gao, S. Synthesis, structure and thermochemical study of a cobalt energetic coordination compound incorporating 3, 5-diamino-1, 2, 4-triazole and pyridine-2, 6-dicarboxylic acid. J. Chem. Thermodyn. 2015, 80, 1-6.
  2. Mohammadi, S.; Khayatian, G. Colorimetric detection of Bi(III) in water and drug samples using pyridine-2, 6-dicarboxylic acid modified silver nanoparticles. Spectrochim. Acta, Part A 2015, 148, 405-411.