Salicylic acid, 99%, Thermo Scientific Chemicals
Salicylic acid, 99%, Thermo Scientific Chemicals
Salicylic acid, 99%, Thermo Scientific Chemicals
Salicylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Salicylic acid, 99%, Thermo Scientific Chemicals
CAS: 69-72-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Catalog number ALFA12253.30
Price (MYR)
265.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
265.00
Add to cart
Specifications
Chemical Name or MaterialSalicylic acid
CAS69-72-7
Melting Point158°C to 160°C
Boiling Point211°C (20mmHg)
Health Hazard 1H302-H318-H361d
View more
Salicylic acid is used as a food preservative as well as in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in dye industry for making azo direct dye and for acidic mordant dyeing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Salicylic acid is used as a food preservative as well as in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in dye industry for making azo direct dye and for acidic mordant dyeing.
Solubility
Soluble in carbon terachloride, benzene, propanol, absolute ethanol, acetone, toluene and ether. Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases, iodine, iron and iron salts.
RUO – Research Use Only
General References:
  1. González-Ramos, K. M.; Fernández-Reyes, B.; Román, F. R.; Hernández-Maldonado, A. J. A hierarchical porous carbon-Mn+[FAU](Mn+ Ni2+ or Cu2+) adsorbent: Synthesis, characterization and adsorption of salicylic acid from water. Microporous Mesoporous Mater. 2014, 200, 225-234.
  2. Ouimet, M. A.; Fogaça, R.; Snyder, S. S.; Sathaye, S.; Catalani, L. H.; Pochan, D. J.; Uhrich, K. E. Poly (anhydride-ester) and Poly (N-vinyl-2-pyrrolidone) Blends: Salicylic Acid-Releasing Blends with Hydrogel-Like Properties that Reduce Inflammation. Macromol. Biosci. 2015, 15 (3), 342-350.