Nonanophenone, 97%, Thermo Scientific Chemicals
Nonanophenone, 97%, Thermo Scientific Chemicals
Nonanophenone, 97%, Thermo Scientific Chemicals
Nonanophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nonanophenone, 97%, Thermo Scientific Chemicals
CAS: 6008-36-2 | C15H22O | 218.34 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12238.0910 g
Catalog number ALFA12238.09
Price (MYR)
444.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
444.00
Add to cart
Specifications
Chemical Name or MaterialNonanophenone
CAS6008-36-2
Melting Point14°C
Recommended StorageAmbient temperatures
Density0.94