3-Octanone, 98%, Thermo Scientific Chemicals
3-Octanone, 98%, Thermo Scientific Chemicals
3-Octanone, 98%, Thermo Scientific Chemicals
3-Octanone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Octanone, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA12113.22100 g
Catalog number ALFA12113.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Octanone
CAS106-68-3
Health Hazard 1H226-H319
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
View more
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Incompatible with strong oxidizing agents, strong reducing agents, strong bases.
RUO – Research Use Only
General References:
  1. Sunada, F. and Heller A. Effects of water, salt water, and silicone overcoating of the TiO2 photocatalyst on the rates and products of photocatalytic oxidation of liquid 3-octanol and 3-octanone. Environ. Sci. Technol. 1998, 32 (2), 282-286.
  2. Raman Ramachandran; Dale M. Norris; Joel K. Phillips; Thomas W. Phillips. Volatiles mediating plant-herbivore-natural enemy interactions: soybean looper frass volatiles, 3-octanone and guaiacol, as kairomones for the parasitoid Microplitis demolitor. J. Agric. Food Chem. 1991, 39 (12), 2310-2317.