1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
CAS: 609-73-4 | C6H4INO2 | 249.01 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12100.1425 g
Catalog number ALFA12100.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Iodo-2-nitrobenzene
CAS609-73-4
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c