1-Phenyl-3-pyrazolidinone, 97%, Thermo Scientific Chemicals
1-Phenyl-3-pyrazolidinone, 97%, Thermo Scientific Chemicals
1-Phenyl-3-pyrazolidinone, 97%, Thermo Scientific Chemicals
1-Phenyl-3-pyrazolidinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Phenyl-3-pyrazolidinone, 97%, Thermo Scientific Chemicals
CAS: 92-43-3 | C9H10N2O | 162.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA12089.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS59377-19-4
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear to turbid colorless to white to yellow
Assay (GC)≥97.5%
Refractive Index1.5925-1.5965 @ 20?C
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.
Solubility
Soluble in water (1:10 hot), methanol (0.1 mg/mL), DMSO (75 mg/mL), and ethanol.
Notes
Store in cool place. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Dennis J. Hlasta.; Francis B. Casey.; Edward W. Ferguson.; Sally J. Gangell.; Martha R. Heimann.; Edward P. Jaeger.; Rudolph K. Kullnig.; Robert J. Gordon. 5-Lipoxygenase inhibitors: the synthesis and structure-activity relationships of a series of 1-phenyl-3-pyrazolidinones. J. Med. Chem. 1991, 34 (5),1560-1570.
  2. Hong-Jun Wang.; Jeffrey Keilman.; Chittari Pabba.; Zhen-Jia Chen.; Brian T. Gregg. Microwave-assisted cross-coupling of 3-chloro-2-pyrazolines and 3-chloro-1-phenyl-1,4,5,6-tetrahydropyridazine with aryl boronic acids. Tetrahedron Lett. 2005, 46 (15),2631-2634.