Pentafluoroiodobenzene, 97%, Thermo Scientific Chemicals
Pentafluoroiodobenzene, 97%, Thermo Scientific Chemicals
Pentafluoroiodobenzene, 97%, Thermo Scientific Chemicals
Pentafluoroiodobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pentafluoroiodobenzene, 97%, Thermo Scientific Chemicals
CAS: 827-15-6 | C6F5I | 293.962 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12000.065 g
Catalog number ALFA12000.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7440-27-9
IUPAC Nameterbium
Molecular FormulaTb
InChI KeyGZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES[Tb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.
Solubility
Insoluble in water.
Notes
Light sensitive. Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. J. M. Birchall.; R. Hazard.; R. N. Haszeldine.; A. W. Wakalski. Polyfluoroarenes. Part VIII. Some homolytic reactions of pentafluoroiodobenzene. J. Chem. Soc. C. 1967, ,47-50.
  2. Ping Cao.; Zheng-Yu Long .;Qing-Yun Chen. Photoinduced Electron-transfer Reaction of Pentafluoroiodobenzene with Alkenes. Molecules. 1997, 2 (1),11-16.