4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals
4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals
4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals
4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals
CAS: 536-40-3 | C7H7ClN2O | 170.60 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11911.065 g
Catalog number ALFA11911.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS591-78-6
IUPAC Namehexan-2-one
Molecular FormulaC6H12O
InChI KeyQQZOPKMRPOGIEB-UHFFFAOYSA-N
SMILESCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.3985-1.4025 @ 20?C