Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals
Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals
Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals
Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals
CAS: 78-94-4 | C4H6O | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ALFA11910.AE
Price (MYR)
902.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
902.00
Add to cart
Specifications
Chemical Name or MaterialMethyl Vinyl Ketone
Name NoteStabilized
CAS78-94-4
Health Hazard 1H225-H300+H310+H330-H314-H317-H335
Health Hazard 2GHS H Statement
H224-H300-H330-H314-H318