4,4'-Stilbenedicarboxylic acid, 96%, Thermo Scientific Chemicals
4,4'-Stilbenedicarboxylic acid, 96%, Thermo Scientific Chemicals
4,4'-Stilbenedicarboxylic acid, 96%, Thermo Scientific Chemicals
4,4'-Stilbenedicarboxylic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Stilbenedicarboxylic acid, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number ALFA11839.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS4025-64-3
IUPAC Name3-(chlorosulfonyl)benzoic acid
Molecular FormulaC7H5ClO4S
InChI KeyLMRKXSDOAFUINK-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
FormPowder
Appearance (Color)White to pale brown
Melting Point (clear melt)125.0-138.0°C