trans-2-Hydroxycinnamic acid, 98+%, Thermo Scientific Chemicals
trans-2-Hydroxycinnamic acid, 98+%, Thermo Scientific Chemicals
trans-2-Hydroxycinnamic acid, 98+%, Thermo Scientific Chemicals
trans-2-Hydroxycinnamic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-2-Hydroxycinnamic acid, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA11740.1425 g
Catalog number ALFA11740.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2345-28-0
IUPAC Namepentadecan-2-one
Molecular FormulaC15H30O
InChI KeyCJPNOLIZCWDHJK-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or fused solid
Assay (GC)≥ 96.0 %
Melting Point37- 41?C
Infrared spectrumConforms
Solution TestColor: Colorless