Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
CAS: 94-18-8 | C14H12O3 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFA11735.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS14405-45-9
IUPAC Nameindium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21InO6
InChI KeyCMGVNFXMZKDHDG-UHFFFAOYSA-N
SMILES[In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)<0.30% Volatiles
FormCrystalline powder
Insoluble material<0.30%