Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
CAS: 94-18-8 | C14H12O3 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALFA11735.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS618-46-2
IUPAC Name3-chlorobenzoyl chloride
Molecular FormulaC7H4Cl2O
InChI KeyWHIHIKVIWVIIER-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration RCOCl>=99.0 %
GC=<0.5 % o-Chlorobenzoyl chloride
Appearance (Color)Clear colorless
Infrared spectrumConforms
Titration RCOOH=<1.0 % (as Chlorobenzoic acid)