Tin(II) oxalate, 98%, Thermo Scientific Chemicals
Tin(II) oxalate, 98%, Thermo Scientific Chemicals
Tin(II) oxalate, 98%, Thermo Scientific Chemicals
Tin(II) oxalate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tin(II) oxalate, 98%, Thermo Scientific Chemicals
CAS: 814-94-8 | C2O4Sn | 206.728 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11689.22100 g
Catalog number ALFA11689.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.02%
Residual halide10mg/l (GC-ECD) heptachlor epoxite
Residue on Evaporation5ppm