Ammonium sulfate, 98+%, Thermo Scientific Chemicals
Ammonium sulfate, 98+%, Thermo Scientific Chemicals
Ammonium sulfate, 98+%, Thermo Scientific Chemicals
Ammonium sulfate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium sulfate, 98+%, Thermo Scientific Chemicals
CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
1000 g
Catalog number ALFA11682.0I
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium sulfate
Melting Point∼280°C (decomposition)
CAS7783-20-2
Recommended StorageAmbient temperatures
RTECS NumberBS4500000
View more
Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.
Solubility
Soluble in water. Insoluble in alcohol and acetone.
Notes
Incompatible with various substances like highly reactive oxidizing agents and alkalis.
RUO – Research Use Only
General References:
  1. Guo, H.; Chen, W.; Sun, X.; Liu, Y. N.; Li, J.; Wang, J. Theranostic magnetoliposomes coated by carboxymethyl dextran with controlled release by low-frequency alternating magnetic field. Carbohydr. Polym. 2015, 118, 209-217.
  2. Carter, D.; Rose, L.; Awobusuyi, T.; Gauthier, M.; Tezel, F. H.; Kruczek, B. Characterization of commercial RO membranes for the concentration of ammonia converted to ammonium sulfate from anaerobic digesters. Desalination 2015, 368, 127-134.