Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
CAS: 141-82-2 | C3H4O4 | 104.061 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA11526.22
Price (MYR)
320.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
320.00
Add to cart
Specifications
Chemical Name or MaterialMalonic acid
CAS141-82-2
Health Hazard 1H302+H332-H315-H318-H335
Health Hazard 2GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
View more
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.
Solubility
Soluble in cold water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Knoevenagel condensation with aldehydes yields substituted acrylic (e.g. cinnamic) acids; review: Org. React., 15, 204 (1967). For an example of the Doebner modification, using pyridine as solvent/base, see: Org. Synth. Coll., 3, 425 (1955); a catalytic amount of piperidine often gives superior results; see, e.g.: Org. Synth. Coll., 4, 327 (1963).
  2. Bew, S. P.; Stephenson, R.; Rouden, J.; Ashford, P. A.; Bourane, M.; Charvet, A.; Dalstein, V. M. D.; Jauseau, R.; Gipson, G. D. H.; Lozano, L. A. M. Bioinspired, Base- and Metal-Free, Mild Decarboxylative Aldol Activation of Malonic Acid Half Thioesters Under Phase-Transfer Reaction Conditions. Adv. Synth. Catal. 2015, 357 (6), 1245-1257.
  3. Nakamura, S.; Sano, M.; Toda, A.; Nakane, D.; Masuda, H. Organocatalytic Enantioselective Decarboxylative Reaction of Malonic Acid Half Thioesters with Cyclic N-Sulfonyl Ketimines by Using N-Heteroarenesulfonyl Cinchona Alkaloid Amides. Chem. Eur. J. 2015, 21 (10), 3929-3932.