1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals
1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals
1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals
1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals
CAS: 295-37-4 | C10H24N4 | 200.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFA11516.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1013-88-3
IUPAC Namediphenylmethanimine
Molecular FormulaC13H11N
InChI KeySXZIXHOMFPUIRK-UHFFFAOYSA-N
SMILESN=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.6170 to 1.6190 (20°C, 589 nm)
View more
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Nitrogen analogue of the crown ethers (see Appendix 2), forming stable complexes with metal ions: Inorg. Chem., 4,1102, 1109 (1965); Can. J. Chem., 48, 1481 (1970). For examples of use as a versatile ligand in coordination chemistry, see: Acc. Chem. Res., 11, 392 (1978); J. Chem. Soc., Chem. Commun., 1322 (1986); 1075 (1987); 156 (1988); Inorg. Chem., 26, 908 (1987); J. Am. Chem. Soc., 110, 3679 (1988).
  2. Complex with iron(II) triflate is an effective epoxidation catalyst: J. Am. Chem. Soc., 113, 7052 (1991).
  3. The Ni(II) complex catalyzes the novel electrochemical reaction of epoxides with carbon dioxide to give good yields of cyclic carbonates: J. Chem. Soc., Chem. Commun., 43 (1995). A similar system catalyzes the electrochemical intramolecular reductive cyclization of a series of o-halogenated aryl alkenes: Tetrahedron Lett., 36, 4429 (1995):
  4. For use in synthesis of molecules with electroactive cavities, see (Ferrocenyl methyl) trimethyl ammonium iodide, 39399.
  5. Misra, A. C.; Luker, K. E.; Durmaz, H.; Luker, G. D.; Lahann, J. CXCR4-Targeted Nanocarriers for Triple Negative Breast Cancers. Biomacromolecules 2015, 16 (8), 2412-2417.