Hypoxanthine, 99%, Thermo Scientific Chemicals
Hypoxanthine, 99%, Thermo Scientific Chemicals
Hypoxanthine, 99%, Thermo Scientific Chemicals
Hypoxanthine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hypoxanthine, 99%, Thermo Scientific Chemicals
CAS: 68-94-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11481.22100 g
Catalog number ALFA11481.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS60583-39-3
IUPAC Namebis((11S,18S,20R,21R,22S)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one) sulfuric acid heptahydrate
Molecular FormulaC46H68N4O19S
InChI KeyPPJIVFYMRNHHTJ-UMRIXEKQSA-N
SMILESO.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<13.0 % (1 g, 105°C, 6 h)
Appearance (Color)White to off-white
Residue after ignition=<0.1 %
Sensitivity(to nitrate) Passes test
View more
Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol A (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol å (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.
Solubility
It does not mix appreciably with water (1:1400). Small octahedra crystals from water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions. Sterilize by filtration through a 0.22 µm filter.
RUO – Research Use Only
General References:
  1. Patrick A. Limbach.; Pamela F. Crain .; James A. McCloskey. the modified nucleosides of RNA. Oxford Journals. 1994, 22, (12), 2183-2196.
  2. James S. Chickos .; William E. Acree Jr. Enthalpies of Sublimation of Organic and Organometallic Compounds. J. Phys. Chem.. 2002, 31, 537.