2-Amino-5-mercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-mercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-mercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-mercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-mercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
CAS: 2349-67-9 | C2H3N3S2 | 133.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA11451.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS82795-51-5
IUPAC Name(2R)-2-amino-4-phenylbutanoic acid
Molecular FormulaC10H13NO2
InChI KeyJTTHKOPSMAVJFE-SECBINFHSA-N
SMILESN[C@H](CCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Optical Rotation-45.0 ± 0.1? (C=1 in 5N HCl)