Bromobenzene, 99%, Thermo Scientific Chemicals
Bromobenzene, 99%, Thermo Scientific Chemicals
Bromobenzene, 99%, Thermo Scientific Chemicals
Bromobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bromobenzene, 99%, Thermo Scientific Chemicals
CAS: 108-86-1 | C6H5Br | 157.01 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11407.36500 g
Catalog number ALFA11407.36
Price (MYR)
378.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
378.00
Add to cart
Specifications
Chemical Name or MaterialBromobenzene
CAS108-86-1
Health Hazard 1H226-H315-H320
Health Hazard 2GHS H Statement
H226-H315
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
View more
Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.
Solubility
Miscible with diethyl ether, alcohol, carbon tetrachloride, chloroform and benzene.
Notes
Store in cool place. Avoid inhalation of vapor or mist. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Man, I. C.; Şoriga, Ş. G.; Pârvulescu, V. Theoretical study of the adsorption of bromobenzene and aniline on Cu2O(1 1 0):CuO and Cu2O(1 1 1):CuO surfaces. Chem. Phys. Lett. 2014, 604, 38-45.
  2. Zhang, X.; Borjian, S.; Baird, M. C. An unusual product distribution from the reaction of Pd(η3-1-PhC3H4)(η5-C5H5) with a deficiency of PPh3 in the presence of bromobenzene. Transition Met. Chem. 2015, 40 (2), 125-129.