Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALFA11379.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS109-65-9
IUPAC Name1-bromobutane
Molecular FormulaC4H9Br
InChI KeyMPPPKRYCTPRNTB-UHFFFAOYSA-N
SMILESCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4380-1.4420 @ 20?C
FormLiquid
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
General References:
  1. Biological buffer.
  2. Lin, L.; Pan, Q. ZnFe2O4@ C/graphene nanocomposites as excellent anode materials for lithium batteries. J. Mater. Chem. A 2015, 3 (4), 1724-1729.
  3. Oishi, M. Enzyme-free and isothermal detection of microRNA based on click-chemical ligation-assisted hybridization coupled with hybridization chain reaction signal amplification. Anal. Bioanal. Chem. 2015, 407 (14), 4165-4172.