Tetra-n-hexylammonium bromide, 98%, Thermo Scientific Chemicals
Tetra-n-hexylammonium bromide, 98%, Thermo Scientific Chemicals
Tetra-n-hexylammonium bromide, 98%, Thermo Scientific Chemicals
Tetra-n-hexylammonium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-hexylammonium bromide, 98%, Thermo Scientific Chemicals
CAS: 4328-13-6 | C24H52BrN | 434.59 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11372.1850 g
Catalog number ALFA11372.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetra-n-hexylammonium bromide
CAS4328-13-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c