o-Xylene, 99%, Thermo Scientific Chemicals
o-Xylene, 99%, Thermo Scientific Chemicals
o-Xylene, 99%, Thermo Scientific Chemicals
o-Xylene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Xylene, 99%, Thermo Scientific Chemicals
CAS: 95-47-6 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
100 mL
500 mL
Catalog number ALFA11358.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Chemical Identifiers
CAS7021-09-2
IUPAC Name2-methoxy-2-phenylacetic acid
Molecular FormulaC9H10O3
InChI KeyDIWVBIXQCNRCFE-UHFFFAOYNA-N
SMILESCOC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Appearance (Color)Off-white to light beige
Appearance (Form)Crystalline powder
Melting point68°C to 72°C
Infrared spectrumConforms
o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.
Solubility
Miscible with ethanol, diethylether. Immiscible with water.
Notes
Incompatible with oxidizing agents and strong acids
RUO – Research Use Only
General References:
  1. Liang, A. D.; Lippard, S. J. Single Turnover Reveals Oxygenated Intermediates in Toluene/o-Xylene Monooxygenase in the Presence of the Native Redox Partners. J. Am. Chem. Soc. 2015, 137 (33), 10520-10523.
  2. Sarafraz-Yazdi, A.; Zendegi-Shiraz, A.; Es’haghi, Z.; Hassanzadeh-Khayyat, M. Synthesis and characterization of composite polymer, polyethylene glycol grafted flower-like cupric nano oxide for solid phase microextraction of ultra-trace levels of benzene, toluene, ethyl benzene and o-xylene in human hair and water samples. J. Chromatogr. A 2015, 1418, 21-28.