Pentadecanolide, 98%, Thermo Scientific Chemicals
Pentadecanolide, 98%, Thermo Scientific Chemicals
Pentadecanolide, 98%, Thermo Scientific Chemicals
Pentadecanolide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pentadecanolide, 98%, Thermo Scientific Chemicals
CAS: 106-02-5 | C15H28O2 | 240.387 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11341.1425 g
Catalog number ALFA11341.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7789-45-9
IUPAC Namecopper(2+) dibromide
Molecular FormulaBr2Cu
InChI KeyQTMDXZNDVAMKGV-UHFFFAOYSA-L
SMILES[Cu++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)Grey to black
Assay (Redox Titration)>98.5 to <101.5%
Solution Test(2% w/v in water): Clear blue solution
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).
Solubility
Soluble in alcohol, dipropylene glycol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Wilberth H-K, et al. Effect of reaction temperature on the physicochemical properties of poly (pentadecanolide) obtained by enzyme-catalyzed ring-opening polymerization. Polymer Bull. 2015, 72(3), 1-12.
  2. Mark B Richardson,; Spencer J Williams. A practical synthesis of long-chain iso-fatty acids (iso-C12-C19) and related natural products. Beilstein Journal of Organic Chemistry. 2013, 9 1807-1812 .