Silicon carbide powder, medium, 120 grit, Thermo Scientific Chemicals
Silicon carbide powder, medium, 120 grit, Thermo Scientific Chemicals
Silicon carbide powder, medium, 120 grit, Thermo Scientific Chemicals
Silicon carbide powder, medium, 120 grit, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silicon carbide powder, medium, 120 grit, Thermo Scientific Chemicals
CAS: 409-21-2 | CSi | 40.10 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11337.36500 g
Catalog number ALFA11337.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS94-36-0
IUPAC Namebenzoyl benzenecarboperoxoate
Molecular FormulaC14H10O4
InChI KeyOMPJBNCRMGITSC-UHFFFAOYSA-N
SMILESO=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Moist granular powder
Appearance (Color)White
Impurity=<0.25 % Inorganic and organic hydrolizable chloride
Titration Iodometric73 to 77 %
Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.
Solubility
Soluble in molten sodium hydroxide, potassium hydroxide and in molten iron.
Notes
Keep it away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Cheng, G.; Chang, T. H.; Qin, Q.; Huang, H.; Zhu, Y. Mechanical Properties of Silicon Carbide Nanowires: Effect of Size-Dependent Defect Density. Electrochem. Commun. 2014, 14 (2), 754-758.
  2. Zhang, X.; Xu, L.; Du, S.; Han, W.; Han, J. Crack-healing behavior of zirconium diboride composite reinforced with silicon carbide whiskers. Scripta Materialia. 2008, 59 (11), 1222-1225.