Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
CAS: 30525-89-4 | CH2O | 30.026 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA11313.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialParaformaldehyde
CAS30525-89-4
Health Hazard 1H228-H302+H332-H315-H317-H318-H335-H351-H500
Health Hazard 2GHS H Statement
H314-H318-H228-H351-H302-H332-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P210-P240-P241-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P370+P378q-P501c
View more
Paraformaldehyde is used as a fumigant, a disinfectant and in the manufacture of synthetic resins like melamine resin, phenol resin and polyacetal resin. It is also used in cell culture. It also serves as a fixative in electron microscopy. It is also used in the preparation of formalin fixatives for tissues or cells.