Melamine, 99%, Thermo Scientific Chemicals
Melamine, 99%, Thermo Scientific Chemicals
Melamine, 99%, Thermo Scientific Chemicals
Melamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Melamine, 99%, Thermo Scientific Chemicals
CAS: 108-78-1 | C3H6N6 | 126.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11295.36500 g
Catalog number ALFA11295.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS6282-02-6
IUPAC NameN-(hydroxymethyl)benzamide
Molecular FormulaC8H9NO2
InChI KeyUOUBPDZUBVJZOQ-UHFFFAOYSA-N
SMILESOCNC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
Melting Point (clear melt)97.0-104.0?C
Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.
Solubility
Slightly soluble in water, hot alcohol, glycerol, glycol and pyridine. Insoluble in ether, benzene and carbon tetrachloride.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Liu, M.; Liu, B.; Shi, L.; Wang, F.; Liang, L.; Sun, J. Melamine-ZnI2 as heterogeneous catalysts for efficient chemical fixation of carbon dioxide to cyclic carbonates. RSC Adv. 2015, 5 (2), 960-966.
  2. Xin, J. Y.; Zhang, L. X.; Chen, D. D.; Lin, K.; Fan, H. C.; Wang,Y.; Xia, C. G. Colorimetric detection of melamine based on methanobactin-mediated synthesis of gold nanoparticles. Food Chem. 2015, 174, 473-479.