(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
CAS: 13822-56-5 | C6H17NO3Si | 179.291 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA11284.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material(3-Aminopropyl)trimethoxysilane
CAS13822-56-5
Health Hazard 1H227-H315-H318
Health Hazard 2GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3P210-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P370+P378q-P501c
View more
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).
Solubility
Miscible with water.
Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.
RUO – Research Use Only
General References:
  1. Schalken, J. R. G.; Creatore, M.; Verhoeven, P.; Stevens, A. mu Plasma Printing Deposition of Amine-Containing Organo-Silane Polymers by Means of 3-Aminopropyl Trimethoxysilane. Nanosci. Nanotechnol. Lett. 2015, 7 (1), 62-66.
  2. Jones, S.; Kolpin, A.; Tsang, S. C. E. Modification of Pd for formic acid decomposition by support grafted functional groups. Catal. Struct. React. 2015, 1 (1), 19-24.