Undecanoic acid, 98%, Thermo Scientific Chemicals
Undecanoic acid, 98%, Thermo Scientific Chemicals
Undecanoic acid, 98%, Thermo Scientific Chemicals
Undecanoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Undecanoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA11244.1850 g
Catalog number ALFA11244.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialUndecanoic acid
CAS112-37-8
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P501c
View more
Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with bases, oxidizing agents, reducing agents. Keep container closed when not in use.
RUO – Research Use Only
General References:
  1. Raluca Voicu; Rabah Boukherroub; Vasiliki Bartzoka; Tim Ward; James T. C. Wojtyk and Danial D. M. Wayner. Formation, Characterization, and Chemistry of Undecanoic Acid-Terminated Silicon Surfaces: Patterning and Immobilization of DNA. Langmuir 2004, 20, (26)
  2. SK Das; AB Banerjee. Effect of undecanoic acid on the production of exocellular lipolytic and keratinolytic enzymes by undecanoic acid-sensitive and -resistant strains of Trichophyton rubrum. Medical Mycology 1982, 20, 179-184.