4-Acetamidophenol, 98%, Thermo Scientific Chemicals
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
CAS: 103-90-2 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1000 g
Catalog number ALFA11240.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS10026-04-7
IUPAC Nametetrachlorosilane
Molecular FormulaCl4Si
InChI KeyFDNAPBUWERUEDA-UHFFFAOYSA-N
SMILESCl[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Copper (Cu)=<0.1 ppm
AppearanceMay develop some turbidity or precipitate
Purity>=99.80 %
Antimony (Sb)=<0.1 ppm
Cobalt (Co)=<0.1 ppm
View more
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.
Solubility
Soluble in alcohol, dimethylformamide, ethylene dichloride, acetone and ethyl acetate. Slightly soluble in water and ether.
Notes
Incompatible with strong oxidizing agents, acids and alkali metals.
RUO – Research Use Only
General References:
  1. Pintal, M.; Kryczka, B.; Marsura, A.; Porwanski, S. Synthesis of bis-cellobiose and bis-glucose derivatives of azacrown macrocycles as hosts in complexes with acetylsalicylic acid and 4-acetamidophenol. Carbohydr. Res. 2014, 386, 18-22.
  2. Albert, A.; Engelhard, C. Chemometric optimization of a low-temperature plasma source design for ambient desorption/ionization mass spectrometry. Spectrochim. Acta, Part B 2015, 105, 109-115.