Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
CAS: 3473-63-0 | C3H8N2O2 | 104.109 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA11158.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFormamidine acetate
CAS3473-63-0
Health Hazard 1H315-H317-H319-H335
Health Hazard 2GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
View more
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
Solubility
Soluble in water and hot alcohol.
Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. More convenient source of formamidine than the corresponding chloride in that it is non-hygroscopic and, being the salt of a weaker acid, can often be used in synthesis without the need to liberate the free base.
  2. Widely used in heterocyclic syntheses, for example of pyrimidine and imidazole derivatives.
  3. Maiden, T. M. M.; Swanson, S.; Procopiou, P. A.; Harrity, J. P. A Mild and Regioselective Route to Functionalized Quinazolines. Chem. Eur. J. 2015, 21 (41), 14342-14346.
  4. Cebrián, C.; Natali, M.; Villa, D.; Panigati, M.; Mauro, M.; D'Alfonso, G.; De Cola, L. Luminescent supramolecular soft nanostructures from amphiphilic dinuclear Re(I) complexes. Nanoscale 2015, 7, 12000-12009.