Trichloroacetic acid, 99%, Thermo Scientific Chemicals
Trichloroacetic acid, 99%, Thermo Scientific Chemicals
Trichloroacetic acid, 99%, Thermo Scientific Chemicals
Trichloroacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trichloroacetic acid, 99%, Thermo Scientific Chemicals
CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1000 g
Catalog number ALFA11156.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS155640-85-0
IUPAC Namecopper(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate)
Molecular FormulaC10H2CuF12O4
InChI KeyHZXGNBMOOYOYIS-PAMPIZDHSA-L
SMILES[Cu++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Iodometric Titration)≥97.5 to ≤102.5% (dry weight basis)
Melting Point100-134?C
Water Content (Karl Fischer Titration)≤8.0%
FormPowder
Appearance (Color)Green
Protein precipitation reagent Trichloroacetic acid is used as a precipitating agent in biochemistry for precipitation of proteins, DNA and RNA. It is an active ingredient used for cosmetic treatments like chemical peels, tattoo removal and the treatment of warts including genital warts. It is also used to determine protein concentration and a decalcifier and fixative in microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Protein precipitation reagent Trichloroacetic acid is used as a precipitating agent in biochemistry for precipitation of proteins, DNA and RNA. It is an active ingredient used for cosmetic treatments like chemical peels, tattoo removal and the treatment of warts including genital warts. It is also used to determine protein concentration and a decalcifier and fixative in microscopy.
Solubility
Soluble in water, alcohol, ethyl ether, benzene, acetone, o-xylene and carbon tetrachloride.
Notes
Moisture sensitive. Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents, strong bases and amines.
RUO – Research Use Only
General References:
  1. Trihaloacetic acids react with aldehydes in DMSO at room temperature to give trihalomethyl carbinols: J. Chem. Soc., Perkin 2, 1247 (1984):
  2. The acid and its Na salt (1:1) in DMF are also effective: Tetrahedron Lett., 33, 3435 (1992). Use of HMPA or DMI extends the scope to less reactive aldehydes: Synthesis, 327 (1990).
  3. Malachová, A.; Štočková, L.; Wakker, A.; Varga, E.; Krska, R.; Michlmayr, H.; Adam, G.; Berthiller, F. Critical evaluation of indirect methods for the determination of deoxynivalenol and its conjugated forms in cereals. Anal. Bioanal. Chem. 2015, 407 (20), 6009-6020.
  4. Young, M. S.; van Tran, K.; Goh, E.; Shia, J. C. A Rapid SPE-Based Analytical Method for UPLC/MS/MS Determination of Aminoglycoside Antibiotic Residues in Bovine Milk, Muscle, and Kidney. J. AOAC Int. 2014, 97 (6), 1737-1741.