Glutaric anhydride, 98%, Thermo Scientific Chemicals
Glutaric anhydride, 98%, Thermo Scientific Chemicals
Glutaric anhydride, 98%, Thermo Scientific Chemicals
Glutaric anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glutaric anhydride, 98%, Thermo Scientific Chemicals
CAS: 108-55-4 | C5H6O3 | 114.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Catalog number ALFA11152.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS617-35-6
IUPAC Nameethyl 2-oxopropanoate
Molecular FormulaC5H8O3
InChI KeyXXRCUYVCPSWGCC-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4035-1.4075 @ 20?C
Assay (GC)≥97.5%
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.
Solubility
Soluble in Ether, dimethyl sulfoxide and methanol and water.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents, acids, bases, alcohols and amines.
RUO – Research Use Only
General References:
  1. Chen, H.; Liu, C.; Chen, D.; Madrid, K.; Peng, S.; Dong, X.; Zhang, M.; Gu, Y. Bacteria-Targeting Conjugates Based on Antimicrobial Peptide for Bacteria Diagnosis and Therapy. Mol. Pharmaceutics 2015, 12 (7), 2505-2516.
  2. Chen, H.; Liu, C.; Chen, D.; Madrid, K.; Peng, S.; Dong, X.; Zhang, M.; Gu, Y. Bacteria-Targeting Conjugates Based on Antimicrobial Peptide for Bacteria Diagnosis and Therapy. Mol. Pharmaceutics 2015, 12 (7), 2505-2516.