Octanoic acid, 98+%, Thermo Scientific Chemicals
Octanoic acid, 98+%, Thermo Scientific Chemicals
Octanoic acid, 98+%, Thermo Scientific Chemicals
Octanoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octanoic acid, 98+%, Thermo Scientific Chemicals
CAS: 124-07-2 | C8H16O2 | 144.214 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11149.AP500 mL
Catalog number ALFA11149.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialOctanoic acid
CAS124-07-2
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.
Solubility
Misicible with alcohol, chloroform, ether, carbon disulfide, petroleum ether and acetonitrile.Slightly miscible with water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Mizrahi, M.; Almogy, G.; Adar, T.; Lysy, J. Erratum: Dumping syndrome following nissen fundoplication in an adult patient diagnosed by continuous online 13C/12C monitoring of 13C-Octanoic acid breath test a case report. BMC Gastroenterol. 2015, 15 (1), 57.
  2. Royce, L. A.; Yoon, J. M.; Chen, Y.; Rickenbach, E.; Shanks, J. V.; Jarboe, L. R. Evolution for exogenous octanoic acid tolerance improves carboxylic acid production and membrane integrity. Metab. Eng. 2015, 29, 180-188.