L-Tyrosine, 99%
L-Tyrosine, 99%
L-Tyrosine, 99%
L-Tyrosine, 99%
Thermo Scientific Chemicals
L-Tyrosine, 99%
CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
100 g
1000 g
Catalog number ALFA11141.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS6203-18-5
IUPAC Name(2E)-3-[4-(dimethylamino)phenyl]prop-2-enal
Molecular FormulaC11H13NO
InChI KeyRUKJCCIJLIMGEP-ONEGZZNKSA-N
SMILESCN(C)C1=CC=C(\C=C\C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with HClO4>=97.5 %
Appearance (Color)Yellow to orange
Melting point137°C to 142°C
Infrared spectrumConforms
L-Tyrosine is used in pharmaceuticals, dietary supplements and food additives. It is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catcholamines. It plays a vital role in photosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Tyrosine is used in pharmaceuticals, dietary supplements and food additives. It is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catcholamines. It plays a vital role in photosynthesis.
Solubility
Soluble in hydrochloric acid. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
  1. Gu, W.; Wang, M.; Mao, X.; Wang, Y.; Li, L.; Xia, W. A facile sensitive l-tyrosine electrochemical sensor based on a coupled CuO/Cu2O nanoparticles and multi-walled carbon nanotubes nanocomposite film. Anal. Methods 2015, 7 (4), 1313-1320.
  2. Hamzehzadeh, S.; Abbasi, M. The influence of 1-butyl-3-methyl-imidazolium bromide on the partitioning of l-tyrosine within the {polyethylene glycol 600+ potassium citrate} aqueous biphasic system at T= 298.15 K. J. Chem. Thermodyn. 2015, 80, 102-111.