Dopamine hydrochloride, 99%, Thermo Scientific Chemicals
Dopamine hydrochloride, 99%, Thermo Scientific Chemicals
Dopamine hydrochloride, 99%, Thermo Scientific Chemicals
Dopamine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dopamine hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 62-31-7 | C8H12ClNO2 | 189.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA11136.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS10401-11-3
IUPAC Name3-ethynylphenol
Molecular FormulaC8H6O
InChI KeyAODMJIOEGCBUQL-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
Appearance (Color)Clear colorless to yellow or brown
FormLiquid
Refractive Index1.5845-1.5895 @ 20?C
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
Solubility
Soluble in water, methanol and dimethyl sulfoxide. Insoluble in ether, petroleum ether, chloroform, benzene and toluene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhai, C.; Li, D.; Li, L.; Sun, F.; Ma, H.; Liu, X. Experimental and theoretical study on the hydrogen bonding between dopamine hydrochloride and N,N-dimethyl formamide. Spectrochim. Acta, Part A. 2015, 145 (7), 500-504.
  2. Gosliga, J. M.; Barter, L. S. Cardiovascular effects of dopamine hydrochloride and phenylephrine hydrochloride in healthy isoflurane-anesthetized New Zealand White rabbits (Oryctolagus cuniculus). Am J Vet Res. 2015, 76 (2), 116-121.